Learn More
Rogo Sampaic™ Glass Coverslips
Preserve and protect specimens on slides.
Brand: Rogo Sampaic™ BPD005
Description
- Available in a range of sizes
- Corrosion-free and resistant to thermal shock
Specifications
Coverslip | |
Soda Lime Glass | |
100/Pk. |
Rectangle and Square | |
24 mm |