Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Gravity chromatography column manufactured from medium wall tubing with 821000 PTFE stopcock
464.00€ - 1110.00€
Specifications
Type | Chromatography Column |
---|---|
For Use With (Application) | Ion exchange chromatography |
Quantity | 1 Each |
USP Type | Type I |
Material | Borosilicate Glass |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
11827433
|
Kimble Chase™
420540-0650 |
500 mm | 64 mm |
769.00€
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
|||||
11837433
|
Kimble Chase™
420540-0660 |
600 mm | 64 mm |
867.00€
Each |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
11847433
|
Kimble Chase™
420540-0750 |
500 mm | 75 mm |
943.00€
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
|||||
11857433
|
Kimble Chase™
420540-0760 |
600 mm | 75 mm |
1110.00€
Each |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
11857423
|
Kimble Chase™
420540-0430 |
300 mm | 38 mm |
464.00€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
13416349
|
Kimble Chase™
420540-0450 |
500 mm | 38 mm |
549.00€
Each |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
11807433
|
Kimble Chase™
420540-0530 |
300 mm | 50 mm |
600.00€
Each |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
11817433
|
Kimble Chase™
420540-0630 |
300 mm | 64 mm |
636.00€
Each |
Estimated Shipment: 05-09-2025
Log in to see stock. |
|||||
Description
- 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
Specifications
Chromatography Column | |
1 Each | |
Borosilicate Glass |
Ion exchange chromatography | |
Type I | |
With fritted disc and PTFE stopcock plug |
Product Title
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug