Learn More
Boric Acid, MP Biomedicals™
Description
Boric acid has a single boron; its salt forms complex equilibria in solution, with the ion B4O7-2 being the predominant species. Manufacturing and industrial uses include: for weatherproofing wood and fireproofing fabrics; as a preservative; manufacturing of cements, crockery, porcelain, enamels, glass, borates, leather, carpets, hats, soaps, artificial gems; in nickeling baths; cosmetics; printing and dyeing, painting; photography; for impregnating wicks; electric condensers; hardening steel. It is also used as an insecticide for cockroaches and black carpet beetles.
Specifications
Specifications
Quantity | 100 g |
Physical Form | White Powder |
Solubility Information | Soluble in water; insoluble in alcohol; freely soluble in boiling water. |
Formula Weight | 61.84 |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Safety and Handling
- Boric Acid, ACS
- Danger
- Reproductive toxicity Category 1B
- H360-May damage fertility or the unborn child.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- MIXTURE LIST-Contains : boric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.