Learn More
Tocris Bioscience™ BOP
Dual α9β1/α4β1 integrin inhibitor; preferentially mobilizes HSCs
358.00€
Specifications
Color | White |
---|---|
Product Type | Inhibitor |
CAS | 1947348-42-6 |
Molecular Formula | C25H28N3O7SNa |
Formula Weight | 537.56 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18767826
|
Tocris Bioscience™
6047/10 |
10 mg |
358.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dual α9β1/α4β1 integrin inhibitor; rapidly and preferentially mobilizes HSCs and progenitors. Augments HSC mobilization in combination with AMD3100 (Cat. No. 3299).Fluorogenic, bright and photostable Fluorescent Conjugate BOP-JF646 (Cat. No. 6997) also available.
High affinity fluorescent derivative R-BC154 (Cat. No. 6048) also available.
Specifications
White | |
1947348-42-6 | |
537.56 | |
Soluble to 100mM in water and to 100mM in DMSO | |
Inhibits integrin |
Inhibitor | |
C25H28N3O7SNa | |
N-(Benzenesulfonyl)-L-prolyl-L-O-(1-pyrrolidinylcarbonyl)tyrosine sodium salt | |
Store at -20°C |
For Research Use Only