Learn More
Syngene Bonded Carbon Filter
AirClean™ Systems bonded carbon filters provide for a predictable and reliable gas phase filtration solution.
Brand: Syngene SYN-ACF12118
This item is not returnable.
View return policy
Description
- Bonded layered carbon filter for organics and inorganics
- Even distribution of impregnates, if applicable
- Multi-layering options for larger application compatibility
Specifications
Carbon | |
Activated Carbon | |
Activated Carbon | |
Chemical extraction |
Airclean | |
Carbon Filter | |
ACF12118 | |
Silver |