Learn More
Bohlender™ BOLA™ Conductors for Magnetic Stirrer Heads
Conductors for Magnetic Stirrer Heads made of borosilicate glass.
965.00€ - 1235.00€
Product Code | Brand | Cone Size | Standard Taper Joint | Thread Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint | Thread Size | Price | Quantity & Availability | |||
17157134
|
Bohlender™
C463-19 |
NS 19/26 | NS 19/26 | Left-handed Thread M 14.5 x 1.5 |
965.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
17167134
|
Bohlender™
C463-45 |
NS 45/40 | NS 45/40 | Left-handed Thread M 21 x 1.75 |
1235.00€
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||
Description
- Borosilicate glass with ground joint and glued upper bearing
- Alternatively without ground joint for fusing on the reactor lid (upper bearing to be glued is included).
Specifications
Borosilicate Glass |