missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP3 Antibody (ANa40), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-1543AF700
This item is not returnable.
View return policy
Description
BNIP3 Monoclonal specifically detects BNIP3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| BNIP3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Q12983 | |
| BNIP3 | |
| Recombinant fragment (Residues 1-163 of Human BNIP3). | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Hypoxia, Phospho Specific, Tumor Suppressors | |
| 664 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| ANa40 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| BNIP3 (ANa40) | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction