missing translation for 'onlineSavingsMsg'
Learn More

BMS 753, Tocris Bioscience™

Product Code. 18055782
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

RARα-selective agonist

RARα-selective agonist (Ki = 2 nM).

Chemical Identifiers

CAS 215307-86-1
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
InChI Key KFBPBWUZXBYJDG-UHFFFAOYSA-N
Synonym d07ytl, bms hplc, 4-1,1,3,3-tetramethyl-2-oxoindene-5-amido benzoic acid, 4-1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl amino benzoic acid, 4-2,3-dihydro-1,1,3,3-tetramethyl-2-oxo-1h-inden-5-yl carbonyl amino benzoic acid
PubChem CID 9884820
IUPAC Name 4-[(1,1,3,3-tetramethyl-2-oxoindene-5-carbonyl)amino]benzoic acid
SMILES CC1(C2=C(C=C(C=C2)C(=O)NC3=CC=C(C=C3)C(=O)O)C(C1=O)(C)C)C

Specifications

Quantity 10 mg
Formula Weight 351.4
Percent Purity >99%
Chemical Name or Material BMS 753
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'