missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP61 Label Printer Kit EU with Workstation PWID Suite

The rugged, thermal transfer BMP61 Label Printer can print sleeves, self-laminating labels, raised profile labels, tags and other identification solutions in a vast range of sizes.

Brand:  Brady™ 198635

Product Code. 16489719

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rugged and reliable, the drop-tested BMP™61 Label Printer is equipped with molded rubber bumpers for extra protection.

  • Fast printing speed
  • Handles die-cut, continuous and custom labels up to 50.8 mm (also in bulk format)
  • Large color touchscreen with indicators allows for easy checking to see which label is installed and how many labels you have left
  • Read spreadsheets directly from external USB-memory and integrate the data into labels ready for printing
  • Wi-Fi version available
  • Compatible with mobile apps- create or load label templates with your smartphone and send them to your printer
  • QWERTY EU keyboard
  • Comes with Brady™ Workstation PWID Suite
Specifications

Specifications

Color LCD
4 in.
13 in.
4 in.
Not Autoclavable
Monochrome
Bulgarian, Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish
BMP61
300 dpi
5°C to 49°C
1.4 kg
2 Year
Thermal Transfer
107 mm
328 mm
106.4 mm
Non-sterile
Printer, USB Cable, Quick Start Guide, Battery Pack, AC Adapter, CD with Driver, Product CD, Brady™ Workstation Product & Wire Identification Suite, Roll of PTL-19-423, M61-R6010 Ribbon
512 MB RAM, 256 MB Flash
33.8 mm/s
USB
3 lb.
Circuit Board and Electrical Labeling, General Identification, Safety Identification
110/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions