Learn More
Brady™ BMP21-LAB Portable Lab Printer Kit UK
The BMP21-LAB Portable Label Printer is an all-in-one labeller for research, academic and clinical labs. Quickly and easily create legible labels for flat, curved or highly textured surfaces that stay stuck for years even when exposed to extreme temperatures and chemicals.
Brand:  Brady™ 134474
Product Code. 15238957
Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 056 260 260.
View alternative products »
Explore more special offers
Description
Description
  • Designed for Laboratory: additional laboratory symbols in font 9 and up, improved barcode quality, ability to preset custom label lengths
  • Labeling: materials available for applications such as cryo vials and slides; Greek symbols commonly used in laboratory, intuitive menu structure for symbols
  • Vial label function automatically cuts the tape according to the size of the vial; Time & Date stamp; Automated serialisation
  • Laboratory specific materials designed to withstand harsh laboratory environments. High quality, long lasting labels that stick and stay stuck
  • Prints durable true-sized 6 – 19 mm wide identification to maximise amount of data. Automatic label formatting for commonly used label types in laboratory
  • Long life rechargeable Lithium-ion battery - accessories sold separately. Battery included in the Starter Kit only
  • 3 line LCD display provides clear visibility
  • 2 year Warranty
Specifications
Specifications
Label Printer Kit EU
Vial and Tube Labels, Slide Labels, Straw Labels, Bottle and Dish Labels
125 mm
50 mm
50 x 125 x 235 mm
Printer, Li-ion Battery, Power Cord, Lanyard, Quick Start Guide, M21-750-7425 label cartridge
10 mm/sec.
5° to 40°C
1 Kit
Monocolor LCD
Thermal Transfer
235 mm
235 x 125 mm
Monochrome
Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Slovak, Slovenian, Spanish, Swedish, Turkish
203 dpi
Laboratory identification
6 AA Batteries, Rechargeable Battery, AC Adaptor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products