Learn More
Blue Marker Antibody (6F4-F6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33376SS
This item is not returnable.
View return policy
Description
Blue Marker Monoclonal specifically detects Blue Marker in Non-species specific samples. It is validated for Western Blot.
Specifications
| Blue Marker | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Detects BLUE prestained molecular weight markers from different vendors, does not cross-react with whole cell proteins of different species, does not interfere with the detection by other antibodies. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| 6F4-F6 | |
| Unconjugated | |
| Blue | |
| Blue dye stained proteins | |
| 0.025 mL | |
| Primary | |
| Non-species specific | |
| Purified |