Learn More
Stuart™ Block Heater Inserts
Product Code. 15592885
Click to view available options
Size:
Double Wide
Triple Wide
Stuart™ Block Heater Inserts offer excellent thermal conductivity; accommodate various tubes and plates
  • Blocks are made of anodized aluminum
  • Features a thermometer hole for easy monitoring of the block's temperature
  • Triple wide Flexi-Block can accommodate multiple size tubes, vials, or cuvettes at once
  • Flexi-blocks use autoclavable high-density ceramic beads
  • Safely heats unskirted PCR plates
  • Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version
Specifications
Material Anodized Aluminum
Type Block Heater Insert
Includes Autoclavable, High-Density Ceramic Beads
Ports Thermometer Hole
Product Line Flexi-block
Size Double Wide