Learn More
Stuart™ Block Heater Inserts
Click to view available options
Size:
Double Wide
Triple Wide
Description
- Blocks are made of anodized aluminum
- Features a thermometer hole for easy monitoring of the block's temperature
- Triple wide Flexi-Block can accommodate multiple size tubes, vials, or cuvettes at once
- Flexi-blocks use autoclavable high-density ceramic beads
- Safely heats unskirted PCR plates
- Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version
Specifications
Specifications
Material | Anodized Aluminum |
Type | Block Heater Insert |
Includes | Autoclavable, High-Density Ceramic Beads |
Ports | Thermometer Hole |
Product Line | Flexi-block |
Size | Triple Wide |