Learn More
BLNK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10855-1-AP-150UL
 View more versions of this product
Product Code. 16827523
  • 410.00€ / 150µL
Description
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
Specifications
Specifications
BLNK
Polyclonal
Unconjugated
BLNK
B cell linker, B cell linker protein, BASH, BLNK, BLNK S, Cytoplasmic adapter protein, LY57, SLP 65, SLP65
Rabbit
Antigen Affinity Chromatography
RUO
17060, 29760
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WV28, Q9QUN3
Blnk
BLNK Fusion Protein Ag1290
150 μL
Primary
Human, Mouse