missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene Blended Carbon Filter
Blended carbon filter for inorganics and organics.
1140.00€ - 1435.00€
Specifications
Filter Type | Carbon |
---|---|
For Use With (Equipment) | Airclean |
Product Type | Carbon Filter |
For Use With (Application) | Chemical extraction |
Color | Silver |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17765944
|
Syngene
SYN-ACF411 |
ACF411 |
1330.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17775944
|
Syngene
SYN-ACF41118 |
ACF41118 |
1140.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17785944
|
Syngene
SYN-ACF411H |
ACF411H |
1410.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17795944
|
Syngene
SYN-ACF414 |
ACF414 |
1320.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17705954
|
Syngene
SYN-ACF41418 |
ACF41418 |
1155.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17715954
|
Syngene
SYN-ACF414H |
ACF414H |
1435.00€
Each |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Spare Part
Specifications
Carbon | |
Carbon Filter | |
Silver |
Airclean | |
Chemical extraction |