missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BLC/CXCL13 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMCXCL13X10
This item is not returnable.
View return policy
Description
CXCL13, also known as BCA-1 and Angie, is a small chemokine strongly expressed in the secondary lymphoid organs. It preferentially promotes the migration of B lymphocytes (both B-1 and B-2) to the follicles of the spleen, lymph nodes, and Peyer's patches. It is produced by macrophages, and in mouse the highest constitutive producers include large peritoneal macrophages, Kuppfer cells and lung macrophages (both CD11c+ and CD11c-). The cellular ligand for CXCL13 is CXCR5 also known as BLR-1 (Burkitt’s Lymphoma Receptor 1). CXCR5 is expressed on B cells, Tfh cells and, to a lower degree, some memory T cell populations. CXCL13 plays an important role in the development of chronic lymphocytic leukemia (CCL). CXCL13 is also a plasma biomarker for germinal center activity and generation of broadly neutralizing antibodies against HIV.Specifications
O55038 | |
2 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Cxcl13 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
1.37 to 1000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
55985 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 5 μL|Serum, 5 μL|Supernatant, 100 μL | |
CXCL13 | |
4 hr 45 min |