Learn More
Brady™ Black 7940 Series Thermal Transfer Printer Ribbon
Ribbon for Brady Printer BBP11 and BBP12
Brand: Brady™ 804458
16.85 EUR valid until 2025-03-09
Use promo code "24130" to get your promotional price.
This item is not returnable.
View return policy
Description
- General purpose, standard performance wax ribbon for paper and temporary labelling
Specifications
Black | |
35 mm | |
Black 7940 Series Thermal Transfer Printer Ribbon | |
1 Ea. |
70 m | |
12.7 mm I.D. | |
Ink side out | |
R7940-35X70/O |