missing translation for 'onlineSavingsMsg'
Learn More

BL 1249, Tocris Bioscience™

Product Code. 18085982
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Putative K2P2.1 (TREK-1) channel opener

Putative potassium channel activator; thought to act as a K2P2.1 (TREK-1) channel opener. Exhibits selectivity for bladder over vascular tissue in vitro and in vivo (EC50 values are 1.26 and 21.0 μM for cultured bladder and aortic tissues respectively).

Chemical Identifiers

CAS 18200-13-0
Molecular Formula C17H17N5
Molecular Weight (g/mol) 291.358
InChI Key YYNRZIFBTOUICE-UHFFFAOYSA-N
Synonym unii-5e7rr1j6tr, 5e7rr1j6tr, 1h-tetrazole, 5-o-5,6,7,8-tetrahydro-1-naphthyl amino phenyl, 1-naphthalenamine, 5,6,7,8-tetrahydro-n-2-1h-tetrazol-5-yl phenyl, 1-naphthalenamine, 5,6,7,8-tetrahydro-n-2-2h-tetrazol-5-yl phenyl, bl hplc, 5,6,7,8-tetrahydro-n-2-2h-tetrazol-5-yl phenyl-1-naphthalenamine, 5,6,7,8-tetrahydro-naphthalen-1-yl-2-1h-tetrazol-5-yl-phenyl-amine, n-2-1h-1,2,3,4-tetrazol-5-yl phenyl-5,6,7,8-tetrahydronaphthalen-1-amine, n-2-2h-1,2,3,4-tetrazol-5-yl phenyl-5,6,7,8-tetrahydronaphthalen-1-amine
PubChem CID 16078951
IUPAC Name N-[2-(2H-tetrazol-5-yl)phenyl]-5,6,7,8-tetrahydronaphthalen-1-amine
SMILES C1CCC2=C(C1)C=CC=C2NC3=CC=CC=C3C4=NNN=N4

Specifications

Quantity 50 mg
Formula Weight 291.35
Percent Purity >99%
Chemical Name or Material BL 1249
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'