missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-toluenesulfonyl)methane, 97%

Product Code. 11302489
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11302489

Brand: Thermo Scientific Alfa Aesar H26617.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15310-28-8 | C15H16O4S2 | 324.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15310-28-8
Molecular Formula C15H16O4S2
Molecular Weight (g/mol) 324.41
MDL Number MFCD00032255
InChI Key XLOXYWLRAKCDQL-UHFFFAOYSA-N
Synonym ditosylmethane, bis-toluene-4-sulfonyl-methane, 1-methyl-4-4-methylbenzenesulfonyl methanesulfonyl benzene, 1,1'-methanediyldisulfonyl bis 4-methylbenzene, acmc-20allu, bis p-tolylsulfonyl methane, bis p-toluenesulfonyl methane, bis-toluene-4-sulfonyl methane, 1,1'-methylenedisulfonyl bis 4-methylbenzene, benzene,1,1'-methylenebis sulfonyl bis 4-methyl
PubChem CID 275690
IUPAC Name 1-methyl-4-[(4-methylphenyl)sulfonylmethylsulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)CS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 137°C to 139°C
Quantity 5 g
Formula Weight 324.42
Percent Purity 97%
Chemical Name or Material Bis(p-toluenesulfonyl)methane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.