missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals

Product Code. 15485796
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944710-34-3
Molecular Formula C20H45NP2
Molecular Weight (g/mol) 361.54
MDL Number MFCD17014018
InChI Key MJYKYLNPIGDVEF-UHFFFAOYSA-N
Synonym bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 51346231
IUPAC Name 2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
SMILES CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C

Specifications

Density 0.911
Odor Unpleasant
Linear Formula ([(CH3)3C]2PCH2CH2)2NH
Refractive Index 1.418
Quantity 5 g
UN Number UN1993
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 361.53
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(di-tert-butylphosphino)ethyl]amine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.