missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carbonyl chloride, 98%

Product Code. 11401708
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11401708

Brand: Thermo Scientific Alfa Aesar A13956.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14002-51-8 | C13H9ClO | 216.66 g/mol

Biphenyl-4-carbonyl chloride was used in the preparation of a novel thiourea compound, N-(6-methyl pyridin-2-yl-carbamothioyl)biphenyl-4-carboxamide. It was also used in the preparation of 5-CF3-oxazole analog, 2-{4-[2-(2-biphenyl-4-yl-5-trifluoromethyl-oxazol-4-yl)-ethoxy]-phenoxy}-2-methyl-propionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4-carbonyl chloride was used in the preparation of a novel thiourea compound, N-(6-methyl pyridin-2-yl-carbamothioyl)biphenyl-4-carboxamide. It was also used in the preparation of 5-CF3-oxazole analog, 2-{4-[2-(2-biphenyl-4-yl-5-trifluoromethyl-oxazol-4-yl)-ethoxy]-phenoxy}-2-methyl-propionic acid.

Solubility
Hydrolysis in water.

Notes
Moisture sensitive. Incompatible with oxidizing agent, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14002-51-8
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000692
InChI Key JPVUWCPKMYXOKW-UHFFFAOYSA-N
Synonym 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride
PubChem CID 84151
IUPAC Name 4-phenylbenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 110°C to 115°C
Boiling Point 160°C (2 mmHg)
Quantity 50 g
UN Number UN3261
Beilstein 472842
Sensitivity Moisture sensitive
Solubility Information Hydrolysis in water.
Formula Weight 216.67
Percent Purity 98%
Chemical Name or Material Biphenyl-4-carbonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.