missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-boronic acid, +97%, Thermo Scientific™

Product Code. 10554964
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4688-76-0
CAS Min % 97.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Molecular Formula C12H11BO2
Linear Formula C6H5C6H4B(OH)2
MDL Number MFCD00136929
Quantity 5 g
Synonym 2-biphenylboronic acid, biphenyl-2-boronic acid, biphenyl-2-ylboronic acid, 2-biphenylboronicacid, 2-phenylphenyl boranediol, 2-phenylphenyl boronic acid, 1,1'-biphenyl-2-ylboronic acid, 2-biphenylylboronic acid, 1,1'-biphenylboronic acid, 2-phenylbenzeneboronic acid
InChI Key HYCYKHYFIWHGEX-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C2=CC=CC=C2)(O)O
IUPAC Name (2-phenylphenyl)boronic acid
Molecular Weight (g/mol) 198.028
PubChem CID 4589187
Formula Weight 198.05
Percent Purity ≥96.0% (HPLC)
Physical Form Powder or Low Melting Solid
Chemical Name or Material Biphenyl-2-boronic acid
Show More Show Less
compliance-icons
Product Identifier
  • Biphenyl-2-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'