missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Biotin-Adrenocorticotropic Hormone (1-39), human

197.00€ - 366.00€

Specifications

Content And Storage Store at -20°C
Format Solid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15450627
View Documents
Thermo Scientific Chemicals
J66673.!D
0.5 mg
197.00€
0.50mg
Please to purchase this item. Need a web account? Register with us today!
15468119
View Documents
Thermo Scientific Chemicals
J66673.M^
1 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Store at -20°C
Solid
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only