Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
Accurate and precise peristaltic pump dispensing.
505.00€ - 713.00€
Product Code | Brand | Increments | Tip Type | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Increments | Tip Type | Volume (Metric) | Price | Quantity & Availability | ||||
16532102
![]() |
Agilent BioTek
1260021 |
1 μL | Steel | 500 nL to 50 μL |
631.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
16582092
|
Agilent BioTek
1260016 |
5 μL | Plastic | 5 to 2500 μL |
505.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||
16592092
![]() |
Agilent BioTek
1260017 |
1 μL | Plastic | 500 nL to 50 μL |
505.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
16542102
![]() |
Agilent BioTek
1260022 |
Not Applicable | Plastic | 8 to 30,000 μL |
713.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
16512102
![]() |
Agilent BioTek
1260019 |
5 μL | Steel | 5 to 2500 μL |
631.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
16572092
|
Agilent BioTek
1260015 |
10 μL | Plastic | 10 to 3000 μL |
505.00€
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||
16502102
![]() |
Agilent BioTek
1260018 |
10 μL | Steel | 10 to 3000 μL |
631.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
16522102
![]() |
Agilent BioTek
1260020 |
10 μL | Plastic, with large bore | 10 to 3000 μL |
505.00€
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
Description
For use with MultiFlo FX Microplate Dispenser RAD peri cassetteSpecifications
MultiFlo FX Microplate Dispenser |
Product Title
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette