missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIO 5192
Highly potent and selective inhibitor of integrin α4β1
407.00€ - 1525.00€
Specifications
Purity | >98% |
---|---|
Product Type | BIO 5192 |
Molecular Weight (g/mol) | 817.78 |
Inhibitors | BIO 5192 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741058
|
Tocris Bioscience™
5051/50 |
50 mg |
1525.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18768188
|
Tocris Bioscience™
5051/10 |
10 mg |
407.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly selective and potent inhibitor of integrin α4β1 (Very Late Antigen-4; VLA-4) (Kd < 10 pM). Selectively binds α4β1 over a range of other integrins (IC50 values are 1.8, 138, 1053, > 500 and > 10,000 nM for α4β1, α9β1, α2β1, α4β7 and αIIbβ3, respectively). Induces a 30-fold increase in mobilization of murine hematopoietic stem and progenitor cells; displays a 3-fold additive effect with AMD 3100 (Cat. No. 3299). Decreases oxidative damage and improves neurological function following spinal cord injury in rats.Specifications
>98% | |
BIO 5192 | |
817.78 | |
BIO 5192 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title