missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Container Connection with Outer Thread

For the easy mounting of thread in the sides of containers.

13.00€

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Used for Easy Mounting of Thread in The Sides of Containers
Color Gray
Product Type Container Connection
View More Specs

Products 1
Product Code Brand Width (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Price Quantity & Availability  
10701611
View Documents
Buerkle™
8530-0012
13.5 mm
13.00€
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • With outer thread
  • Complete with EPDM seals
Specifications

Specifications

Autoclavable
Used for Easy Mounting of Thread in The Sides of Containers
Container Connection
Polypropylene
Gray
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.