missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, Reagent Grade, 99%, Honeywell™

Product Code. 15607710
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-47-0
Molecular Formula C7H5N
Molecular Weight (g/mol) 103.124
MDL Number MFCD00001770
InChI Key JFDZBHWFFUWGJE-UHFFFAOYSA-N
Synonym phenyl cyanide, cyanobenzene, benzenenitrile, benzoic acid nitrile, benzene, cyano, benzenecarbonitrile, phenylcyanide, fenylkyanid, fenylkyanid czech, benzonitril
PubChem CID 7505
ChEBI CHEBI:27991
IUPAC Name benzonitrile
SMILES C1=CC=C(C=C1)C#N

Specifications

Melting Point -13°C
Color Colorless
Boiling Point 191°C
Flash Point 70°C (158°F)
Packaging Glass Bottle
Linear Formula C6H5CN
Quantity 1 L
UN Number UN2224
Beilstein 506893
Vapor Density 3.56 (Air = 1.0)
Formula Weight 103.12g/mol
Vapor Pressure 1 hPa at 20°C
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'