missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Switch-Grid™ Test Tube Rack Grids
Replacement grids
Brand: Bel-Art™ F18745-1600
This item is not returnable.
View return policy
Description
Bel-Art™ Scienceware™ Switch-Grid™ Test Tube Rack Grids
- Chemical resistant polyoxymethylene plastic
- Can be cleaned with mild detergent, ethanol or isopropyl alcohol
- Autoclavable at121°C (250°F)
Specifications
Grid Set | |
Blue | |
Plastic |
6 x 5 | |
Tubes (13 to 16 mm) | |
60 |