missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture Orbital Shaker Platform

Turns a standard magnetic stirrer into an orbital shaking platform saving the expense and space requirements of additional lab equipment.

Brand:  Bel-Art™ F37041-0000

Product Code. 12338059

  • 460.84€ / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perfect for dishes, flasks, and trays, it is also ideal for Western Blot processing.

  • Shake and mix; rotating orbit diameter of 21.7mm (7/8 in.)
  • Shaker adjustment is controlled by setting the magnetic stirrer to the desired speed
  • Platform protects the operating mechanism from spills
  • Non-skid surface prevents containers from sliding
  • Base incorporates built-in hand holds for lifting and carrying
  • Supports up to 6.8 kg (15 lb.)
  • Dimensions: 8.9 x 8.9 x 2.1 in.
  • Weight: 2.2 lb.

U.S. Patent 5,409,312.

Specifications

Specifications

Orbital Platform Shaker
2.1 in.
8.9 in.
8.9 in.
2.25 lb.
8.9 x 8.9 x 2.1 in. (22.5 x 22.5 x 5.3 cm)
5.3 cm
22.5 cm
22.5 cm
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.