Learn More
behr Labor-Technik™ behrotest™ Reflux Condenser Allhin
Prevent solvents from boiling away. behr Labor-Technik™ behrotest™Reflux Condenser Allhin is used to prevent solvent from boiling away. As vapor from the boiling solvent rises into inside column of the reflux condenser, it is cooled by the jacket of water on the outside and condenses.
Brand:  behr Labor-Technik™ B00217957
Product Code. 10004771
  • 227.00€ / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With oval bulbs
Specifications
Specifications
2.7 L
0.48 m
0.045 m
Width: 0.125m, Height: 0.045m, Depth: 0.48m
0.125 m