Learn More
BD Green Nucleic Acid Stain
Product Code. 15814539
Click to view available options
Quantity:
0.1 mL
0.5 mL
Unit Size:
Each
Product Code. 15814539
Brand: BD 565802
Useful for discrimination of viable from nonviable cells, DNA content analysis in fixed cells, or nuclear counterstaining for fixed cells in immunofluorescence or flow cytometry applications. BD Via-Probe™ Green Nucleic Acid Stain brightly stains nonviable or fixed cells with permeable membranes.
  • Green Nucleic Acid Stain has an excitation maximum of 503nm, and is best excited by the blue laser
  • It has an emission maximum of 526nm
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.5 mL
For Use With (Application)
  • Immunofluorescence
  • Immunocytochemistry
  • Flow cytometry
  • Intracellular staining (flow cytometry)
Color Green
Product Type Green Nucleic Acid Stain