Learn More
BD Urine Preservative Transport
Extends range of time and temperature conditions for CT/GC specimen storage and transport
Brand: BD 441362
Includes: Each Qx UPT Kit contains 100 Qx UPT tubes and 100 transfer pipettes. Each UPT contains 50μL of NAP Guard™ (≥ 742.5mM K2EDTA).
Description
Qx Urine Preservative Transport
- Allows for an extended range of time and temperature conditions for specimen storage and transport
- Designed to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae
- For male and female urine specimens from symptomatic and asymptomatic individuals
- For use with the BD ProbeTec™ CT Qx and GC Qx Amplified DNA Assays on the BD Viper™ System in extracted mode
Specifications
Urine Preservative Tube | |
NAP Guard™, 50μL | |
For BD ProbeTec™ Chlamydia trachomatis and Neisseria gonorrhoeae Qx amplified DNA Assays, to preserve and transport Chlamydia trachomatis and Neisseria gonorrhoeae in male and female Urine specimens | |
Each Qx UPT kit contains 100 Qx UPT tubes and 100 transfer pipettes, each UPT contains 50μL of NAP Guard™ |