missing translation for 'onlineSavingsMsg'
Learn More

BD Port-A-Cul™ Anaerobic Collection and Transport Systems

Versatile—use for fluid, tissue, or swab specimens

Brand:  BD 221602

 View more versions of this product

Product Code. 17182846

  • 690.00€ / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Collect and maintain specimens at 20° to 25°C for up to 72 hours
  • Ensure aerobic, anaerobic, microaerophilic or facultative microorganism viability during transport from patient to laboratory (including Campylobacter jejuni)
  • Reduced medium formulated to protect oxygen-sensitive microbes
  • Media contain an indicator that continuously monitors the reduced media status
  • Collect specimen on a swab, as a fluid, or as a tissue specimen
  • Available as tube system (capacity 11mL), jar system (capacity 20mL), or vial system (capacity 5mL)
  • Sterile Package contains either one tube and two polyester-tipped wooden swabs, one vial or one jar
  • Screw-cap jar is 3 in. tall and has a 1 in.-wide mouth; not for use with fluids
  • Fluid Collection Kit contains one sterile-packaged vial containing media and a 5cc syringe with a 22-gauge needle and Luer-Lok™ closure

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Port-A-Cul™ Transport Jar
To maintain the viability of anaerobic, facultative and aerobic microorganisms during transport from the patient to the laboratory
20°C to 25°C
Sterile
10 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.