Learn More
BD Mouse Th17/Treg Phenotyping Kit
Simultaneously view TH17 and regulatory T cells in mouse model systems
Brand:  BD 560767
665.11 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15810479
  • 670.00€ / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Includes: CD4 PerCP-Cy5.5, IL-17A PE, Foxp3 Alexa Fluor 647 antibody cocktail, Foxp3 Buffer Set, BD GolgiStop Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
For research use only. Not for diagnostic or therapeutic procedures.