missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Oxford Medium Base

Product Code. 11772734
Click to view available options
:
Oxford Medium Base; 500g
This item is not returnable. View return policy

Product Code. 11772734

Brand: BD 222530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with Modified Oxford Antimicrobic Supplement or Oxford Antimicrobic Supplement to isolate L. monocytogenes

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, ISO, SMD, USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type Oxford Medium Base
Product Type Oxford Medium Base
Formulation Pancreatic digest of Casein 8.9g/L, Proteose Peptone 4.4g/L, Yeast extract 4.4g/L, Tryptic digest of Beef heart 2.7g/L, Starch 0.9g/L, Sodium Chloride 4.4g/L, Esculin 1g/L, Ferric Ammonium Citrate 0.5g/L, Lithium Chloride 15g/L, Agar 15.3g/L
For Use With (Application) For isolating and differentiating Listeria monocytogenes
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.