missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse TNF; Flex Set, Bead C8

For detection of mouse tumor necrosis factor in serum and cell culture supernatant samples

Brand:  BD 558299


Product Code. 10678954

  • 560.00€ / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket

Includes: Mouse TNF; Standard; Mouse TNF; PE Detection Reagent; Mouse TNF; Capture Bead C8
This item is not returnable. View return policy

Description

Description

The BD CBA Mouse TNF Flex Set is a bead-based immunoassay capable of measuring mouse tumor necrosis factor (TNF) in serum and cell culture supernatant samples. Mouse and rat reactivity was determined by testing samples with the BD CBA Mouse TNF Flex Set. For quantitation of rat TNF, we recommend the BD™ CBA Rat TNF Flex Set (Cat. No. 558309).

  • Reactivity: Mouse and Rat
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Applications:

Flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNF
Mouse TNF, standard, Mouse TNF, PE detection reagent, Mouse TNF, capture bead C8
Serum, Cell Culture Supernatant
Flow Cytometry
Mouse
100 Tests
4°C
Mouse TNF flex set (bead C8)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MS TNF CBA FLEX SET 100TST C8

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isoyhiazolin-3-one

SDS
Documents

Documents

Certificates
Promotions

Promotions

Research Use Only