Learn More
Cytometric Bead Array (CBA) Mouse/Rat Soluble Protein Master Buffer Kit, 100 tests, BD
Employs particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations
Brand:  BD Biosciences 558266
Product Code. 10318813
  • 635.00€ / Each
Explore more special offers
Includes: Assay Diluent, Capture Bead Diluent, Detection Reagent Diluent, Wash Buffer, Instrument Setup Beads (A1, A9, F1, F9), PE Instrument Setup Bead F1, PE Positive Control Detector
Description
Description
This kit contains all supporting reagents necessary to perform an assay using BD™ CBA Mouse or Rat Soluble Protein Flex Set. Buffers and instrument setup reagents are optimized for analysis of analytes in tissue culture supernatants, plasma and serum samples.
  • Analyte working assay range: 10 to 2500pg/mL (mouse), 40 to 10000pg/mL (rat)
  • Should not be used with any non-Mouse or Rat Soluble Protein Flex Sets
  • Available in pack sizes of 100 and 500 tests
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2°C to 8°C
100
1 bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 130mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1