missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array (CBA) Mouse KC; Flex Set, Bead A9

For detecting mouse keratinocyte-derived chemokine in serum and cell culture samples

Brand:  BD 558340

Product Code. 15839318

  • 585.00€ / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket

Includes: Mouse KC; Standard; Mouse KC; PE Detection Reagent; Mouse KC; Capture Bead A9
This item is not returnable. View return policy

Description

Description

The BD CBA Mouse KC Flex Set is a bead-based immunoassay capable of measuring mouse keratinocyte-derived chemokine (KC), a mouse functional homolog of human GRO-?, in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse KC Flex Set.

  • Reactivity: Mouse
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

TRUSTED_SUSTAINABILITY
Specifications

Specifications

KC
Mouse KC, standard, Mouse KC, PE detection reagent, Mouse KC, capture bead A9
Serum, Cell Culture Supernatant
Flow Cytometry
Mouse
100 Tests
4°C
Mouse KC flex set (bead A9)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Research Use Only