Learn More
BD Cytometric Bead Array (CBA) Human IgE; Flex Set, Bead E9
Bead-based immunoassay for measuring human total immunoglobulin E (IgE) in serum and plasma samples
Brand: BD 558682
Includes: Human IgE, Standard (two vials), Human total IgE PE Detection Reagent (one vial), Human total IgE Capture Bead E9 (one vial)
Description
- Human reactivity was determined by testing samples with IgE Flex Set
- Theoretical limit of detection is 0.07ng/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
IgE | |
Human IgE, standard (2 vials), Human total IgE PE detection reagent (one vial), Human total IgE capture bead E9 (one vial) | |
4°C | |
Human IgE flex set (bead E9) |
Human | |
100 Tests | |
Flow Cytometry |
Research use only