Learn More
BD Cytometric Bead Array (CBA) Human CD14; Flex Set, Bead D7
Determines human reactivity by testing samples with BD CBA Human Soluble CD14 Flex Set
Brand:  BD 560418
Product Code. 15890179
  • 835.00€ / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Alert:
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Includes: Human CD14; Standard; Human CD14; PE Detection Reagent; Human CD14; Capture Bead D7
Description
Description
The BD CBA Human CD14 Flex Set is a bead-based immunoassay capable of measuring human soluble CD14 in cell culture supernatant samples
  • Reactivity: QC Testing: Human
  • Bead Position: D7
  • Limit of detection: 9.2pg/mL
With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), flow cytometer, and FCAP Array™software
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD14
Human CD14, standard, Human CD14, PE detection reagent, Human CD14, capture bead D7
4°C
Human soluble CD14 flex set (bead D7)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Research use only