missing translation for 'onlineSavingsMsg'
Learn More

BD AEC Substrate Kit

Substrate for peroxidase and is a widely used chromogen for immunohistochemical staining and immunoblotting

335.00€ - 1160.00€

Specifications

Chromagen Type AEC
For Use With (Application) Immunohistochemical staining and immunoblotting, ELISPOT
Substrate Type AEC Substrate
pH 7.2
Storage Requirements 4°C
View More Specs

Alert:

Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.


Includes: AEC Buffer and AEC Chromogen

Products 2
Product Code Brand Chromagen Quantity Includes Price Quantity & Availability  
Product Code Brand Chromagen Quantity Includes Price Quantity & Availability  
15826478
View Documents Promotion Details
BD
551015
1.5mL AEC buffer and AEC chromogen
1160.00€
1 set
Please to purchase this item. Need a web account? Register with us today!
15836738
View Documents Promotion Details
BD
551951
3mL 10 plates, AEC buffer and AEC chromogen
335.00€
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunohistochemistry, ELISPOT

Specifications

Specifications

AEC
AEC Substrate
4°C
Immunohistochemical staining and immunoblotting, ELISPOT
7.2
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • AEC Substrate Kit

Signal Word
  • Danger||Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
  • H371-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol, N,N-dimethyl formamide, 9-ethylcarbazol-3-ylamine

SDS
Documents

Documents

Product Certifications

Research Use Only