missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-X Mouse, Clone: BX006 + 2H12, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 598-MSM3-P1
This item is not returnable.
View return policy
Specifications
| Bcl-X | |
| Cytoplasmic, cell/nuclear membrane | |
| BX006 + 2H12 | |
| Unconjugated | |
| NP_612815 | |
| Mouse | |
| 27kDa | |
| Protein A/G Purified | |
| RUO | |
| 598 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| BCL2L1 | |
| A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein (BX006 and 2H12) | |
| Jurkat, K562, HL-60, or HeLa Cells. Reed-Sternberg cells in Hodgkin's lymphoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction