Learn More
Tocris Bioscience™ BCH
LAT1 inhibitor
Brand:  Tocris Bioscience™ 5027/50
Product Code. 18738547
  • 104.00€ / 50mg
Explore more special offers
Description
Description
BCH is an inhibitor of L-type amino acid transporter LAT1. Suppresses growth and induces apoptosis via activation of caspases in KB, Saos2 and C6 cancer cell lines.
Specifications
Specifications
50 mg
BCH
155.2
BCH