Learn More
Bayonet 1140 A2B2E2K2HgP3 R D Combination Filter
Brand:  Draeger Safety 6738797
Additional Details : Weight : 0.39600kg
Product Code. 10311245
  • 56.05€ / Each
In Stock
to see stock.
Specifications
EN12941 and EN12942
Combination Filter
1 Ea.
1140 A2B2E2K2HgP3 R D
Can be Used as Filter TH/m3 A2B2E2K2 Hg PSL in the Powered Air Purifying Respirator Dräger X-plore™