missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAY 60-6583, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent adenosine A2B receptor agonist (EC50 = 2.83 nM for murine A2B receptor). Displays selectivity for A2B over A1, A2A and A3 receptors. Decreases fMLP-induced superoxide production in neutrophils at low concentrations (1-10 nM). Cardioprotective; attenuates infarct size in a mouse model of myocardial ischemia. Exhibits ERK1/2-biased agonism.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 379.44 |
Percent Purity | >98% |
Chemical Name or Material | BAY 60-6583 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'