missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bay 11-7085, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Irreversible inhibitor of TNF-α-stimulated IκBα phosphorylation (IC50 ∽ 10 μM); leads to decreased NF-κB and subsequent decreased expression of adhesion molecules. Also reversibly activates MAP kinases and stimulates apoptosis. Anti-inflammatory in vivo.
Specifications
Specifications
CAS | 196309-76-9 |
Molecular Formula | C13H15NO2S |
Quantity | 10 mg |
Synonym | e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile |
InChI Key | VHKZGNPOHPFPER-ONNFQVAWSA-N |
SMILES | CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N |
IUPAC Name | (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile |
Molecular Weight (g/mol) | 249.328 |
PubChem CID | 5353432 |
Formula Weight | 249.33 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'