missing translation for 'onlineSavingsMsg'
Learn More

Batimastat, Tocris Bioscience™

Product Code. 18063792
Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, broad spectrum MMP inhibitor

Potent, broad spectrum MMP inhibitor (IC50 values are 3, 4, 4, 6 and 20 nM for MMP -1, -2, -9, -7 and -3 respectively). Exhibits antiproliferative, anti-invasive and antimetastatic activity in human ovarian carcinoma xenografts in vivo.

Chemical Identifiers

CAS 130370-60-4
Molecular Formula C23H31N3O4S2
Molecular Weight (g/mol) 477.638
InChI Key XFILPEOLDIKJHX-QYZOEREBSA-N
Synonym batimastat, batimastat bb-94, unii-bk349f52c9, butanediamide, n4-hydroxy-n1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*, 2s,3r-5-methyl-3-alphas-alpha-methylcarbamoyl phenethyl carbamoyl-2-2-thienylthio methyl hexanohydroxamic acid, 4-n-hydroxyamino-2r-isobutyl-2s-2-thienylthiomethyl succinyl-l-phenylalanine-n-methylamide, 2r-n-1s-1-benzyl-2-methylamino-2-oxo-ethyl-2-1s-2-hydroxyamino-2-oxo-1-2-thienylsulfanylmethyl ethyl-4-methyl-pentanamide, 2r,3s-n-hydroxy-n'-1s-1-methylcarbamoyl-2-phenylethyl-2-2-methylpropyl-3-thiophen-2-ylsulfanyl methyl butanediamide, butanediamide, n sup 4-hydroxy-n sup 1-2-methylamino-2-oxo-1-phenylmethyl ethyl-2-2-methylpropyl-3-2-thienylthio methyl-, 2r-1 s* ,2r*,3s*
PubChem CID 5362422
IUPAC Name (2S,3R)-N-hydroxy-N'-[(2S)-1-(methylamino)-1-oxo-3-phenylpropan-2-yl]-3-(2-methylpropyl)-2-(thiophen-2-ylsulfanylmethyl)butanediamide
SMILES CC(C)CC(C(CSC1=CC=CS1)C(=O)NO)C(=O)NC(CC2=CC=CC=C2)C(=O)NC

Specifications

Quantity 1 mg
Formula Weight 477.64
Percent Purity >99%
Chemical Name or Material Batimastat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.