Learn More
Invitrogen™ Basic Security Module, FlexMAP™ 3D™ System
Product Code. 10385119
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. 10385119
Brand: Invitrogen™ FM3D0BS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The basic security module provides security by allowing an administrator to set up user access levels.
The basic security module provides security by allowing an administrator to set up user access levels. This module is designed for the FlexMAP™ 3D™ system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FlexMAP™ 3 D™ System
Product Line Luminex
Quantity 1 each
Content And Storage Store at room temperature
Shipping Condition Room Temperature
Software Security