Learn More
Thermo Scientific™ Barnstead™ MegaPure™ ACS
Control the operation of your Thermo Scientific™ Barnstead™ Mega-Pure™ Glass Still with this automatic collection system, which collects up to 45L of water.
Brand:  Thermo Scientific™ 400634
 View more versions of this product
Product Code. 12743848
  • 522.60€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Control the operation of your glass still with the Thermo Scientific™ Barnstead™ MegaPure™ ACS (Automatic Collection System), which collects up to 45L of water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACS Wall mounting bracket
Automatic Collection System (ACS)
Wall Bracket