missing translation for 'onlineSavingsMsg'
Learn More

Barbituric acid, 99+%

Product Code. 10656404 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10656404

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barbituric acid, CAS # 67-52-7, (synonym: malonylurea) is the parent compound used in the production of barbiturates that act as sedative drugs. It is also used extensively in the manufacturing of plastics, textiles, and polymers. | CAS: 67-52-7 | C4H4N2O3 | 128.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Barbituric acid is a white, odorless, water-soluble crystalline powder
• It is a strong acid in an aqueous medium


Applications

• Barbituric acid uses the Knoevenegal condensation reaction to be converted into barbiturate drugs that acts as central nervous system depressants
• It is an active ingredient in the synthesis of Vitamin B12
• It is also used in electrochemical oxidation of iodine using cyclic voltammetry and controlled potential coulometry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-52-7
Molecular Formula C4H4N2O3
Molecular Weight (g/mol) 128.09
MDL Number MFCD00006666
InChI Key HNYOPLTXPVRDBG-UHFFFAOYSA-N
Synonym malonylurea, pyrimidine-2,4,6 1h,3h,5h-trione, barbiturate, 6-hydroxyuracil, 2,4,6 1h,3h,5h-pyrimidinetrione, malonylharnstoff, 2,4,6-pyrimidinetrione, barbituric acid, 2,4,6-trihydroxypyrimidine, pyrimidinetrione
PubChem CID 6211
ChEBI CHEBI:16294
IUPAC Name 1,3-diazinane-2,4,6-trione
SMILES O=C1CC(=O)NC(=O)N1

Specifications

Assay Percent Range 99+%
Merck Index 15, 958
Formula Weight 128.09
Quantity 500 g
Packaging Plastic bottle
Chemical Name or Material Barbituric acid, 99+%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.